missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DETACHaBEAD™ CD19 Kit DFS Item

B cells isolated with Dynabeads™ CD19 and released with DETACHaBEAD™ CD19 are pure, viable, phenotypically unaltered and unactivated.

Supplier:  Invitrogen™ 12506D

Catalog No. 12506D


Only null left
Explore available promotions

Includes: DETACHaBEAD CD19, polyclonal anti-Fab to release cells from Dynabeads CD19 (Pan B)

Description

Description

B cells isolated with Dynabeads™ CD19 and released with DETACHaBEAD™ CD19 are pure, viable, phenotypically unaltered and unactivated. Cells are suitable for applications such as further sorting, culturing studies, and flow cytometry.

This Product Contains: DETACHaBEAD™ CD19, polyclonal anti-Fab to release cells from Dynabeads™ CD19 (Pan B). For use with cells isolated using Dynabeads™ CD19

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

B Cells
Antibody Coated Bead
>95%
Research Grade
Human
Room Temperature
Human
Store in refrigerator (2–8°C).
Positive Isolation
DETACHaBEAD, Dynabeads
5 mL
PBMC, Blood
1 x 107 PBMCs per isolation
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.