missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ dNTP Mix (10 mM ea)

Catalog No. 18427013
Change view
Click to view available options
Quantity:
100 μL
1 mL
5 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18427013 100 μL
18427089 5 x 1 mL
18427088 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18427013 Supplier Invitrogen™ Supplier No. 18427013

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen 10 mM dNTP Mix includes four nucleotides (dATP, dCTP, dGTP, dTTP) in 0.6 mM Tris-HCl. Each nucleotide is at a concentration of 10 mM and this mix is suitable for use in a wide variety of molecular processes.

Invitrogen 10 mM dNTP Mix includes four nucleotides (dATP, dCTP, dGTP, dTTP) in 0.6 mM Tris-HCl. Each nucleotide is at a concentration of 10 mM and this mix is suitable for use in a wide variety of molecular processes including: polymerase chain reaction (PCR), sequencing, fill-in reactions, nick translation, cDNA synthesis, and TdT-tailing reactions.

Features include:
• Chemically synthesized
• >99% purity confirmed by HPLC
• Stable for 2 years at –20°C
• Free from qPCR, PCR, reverse transcription inhibitors
• Free of DNases and RNases
• Free of human and E. coli DNA

Applications
• qPCR, RT-qPCR
• PCR, RT-PCR, cDNA synthesis
• High-fidelity and long-range PCR
• Isothermal amplification
• DNA labeling
• Cloning
• Sanger and next-generation sequencing (NGS)

Thermo Fisher Scientific offers a wide variety of dNTPs in different concentrations, volumes, and formats.

Order Info

Approved for shipment on Wet or Dry Ice

Specifications

Concentration 10 mM
Content And Storage dNTP Mix, contains 10 mM each of dATP, dCTP, dGTP, and dTTP (100 μL)

Store at –20°C.
Form Liquid
Label or Dye Unlabeled
Quantity 100 μL
Shipping Condition Approved for shipment on Wet or Dry Ice
For Use With (Application) Standard PCR, RT-PCR, Real Time PCR (qPCR)
Which components of the SuperScript III First Strand Synthesis System for RT-PCR are available for purchase separately?

The following components are available as stand-alone items:

- Superscript III Reverse Transcriptase (Cat. Nos. 18080093, 18080044, 18080085)
- Oligo (dT)20 Primer (Cat. No. 18418020)
- Random hexamers (Cat. No. 48190011)
- 10 mM dNTP Mix (Cat. Nos. 18427013, 18427088)
- RNAseOUT Recombinant Ribonuclease Inhibitor (Cat. No. 10777019)
- E. coli RNAse H (Cat. Nos. 18021014, 18021071)

Why do some dNTP mixes contain dTTP and others TTP?

Thymine is normally found as a deoxyribose. Thus, TTP and dTTP are essentially the same thing. Both types of mixes can be used in PCR reactions

Why does the annealing temperature have to be above 55 degrees C, when using the GeneAmp PCR Carryover Prevention kit (Cat. No. N808-0068) or any kit containing AmpErase UNG (Cat. No. N808-0096)?

Uracil-N-glycosylase (UNG) has activity in the temperature range of 4 degrees C to 55 degrees C, and is not completely inactivated by incubation at 95 degrees C. It is recommended that the annealing temperature be at or above 55 degrees C when using this enzyme with any of our PCR or Sequence Detection Systems kits to avoid possible degradation of newly synthesized dU-containing PCR products by residual UNG activity. For this reason, it is also recommended that the post-PCR soak be maintained at 72 degrees C (instead of 4 degrees C) if analysis of the PCR product will be performed using a gel-based method.

What concentration of dNTPs is typically used in a PCR amplification?

The four dNTPs (dATP, dCTP, dTTP, dGTP) are used at a final concentration of 200 µM each or a total of 0.8 mM in standard amplifications. For PCR amplifications that require high fidelity, lower dNTP concentrations (on the order of 10-50 µM) work optimally.

Describe the amplified product produced by the control reagents in the GeneAmp Gold PCR Reagent Kit with Multiplex Control (Cat. No. 4312778).

The control primers generate a 300 and a 500 bp amplicon from the lambda control DNA. The Lambda Multiplex Control primers, as well as the Lambda Control DNA are provided in this kit.

For Research Use Only. Not for use in diagnostic procedures.