Learn More
Invitrogen™ DNA Polymerase I, Large (Klenow) Fragment
Description
Includes
DNA Polymerase I, Large (Klenow) Fragment is supplied with a vial of 10X REact™ 2 Buffer [500mM Tris-HCl (pH 8.0), 100mM MgCl2, 500mM NaCl], vial of dilution buffer.
Applications:
Fill-in of 5' overhangs . Synthesis of probes by random primers labeling method. Sequencing single- and double-stranded DNA. Site-directed mutagenesis.
Source:
Purified from E. coli expressing the Klenow fragment on a plasmid.
Performance and Quality Testing:
SDS-PAGE purity; single-stranded and double-stranded endodeoxyribonuclease and self-priming assays; performance evaluated in fill-in reaction.
Unit Definition:
One unit incorporates 10nMol of total deoxyribonucleotide into acid-precipitable material in 30 min. at 37°C using a template•primer.
Array CGH, Cloning, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Restriction Enzyme Cloning
Order Info
Shipping Condition: Approved for shipment on Wet or Dry Ice
Specifications
Specifications
| Content And Storage | DNA Polymerase I, Large (Klenow) Fragment is supplied with a vial of 10X REact™ 2 Buffer [500 mM Tris-HCl (pH 8.0), 100 mM MgCl2 , 500 mM NaCl], vial of dilution buffer. Store at -20°C. |
| Description | DNA Polymerase |
| Polymerase | DNA Polymerase I |
| Exonuclease Activity | 3' to 5' |
| Quantity | 100 Units |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
| Hot Start | No |
For Research Use Only. Not for use in diagnostic procedures.