missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA Polymerase I

Catalog No. p-4921658
Encompass
Change view
Click to view available options
Quantity:
1000 U
250 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18010017 250 U
18010025 1000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18010017 Supplier Invitrogen™ Supplier No. 18010017
Only null left

Includes

1000 units of DNA polymerase in a storage buffer of 50mM potassium phosphate (pH 7.0), 100mM KCl, 1mM DTT, and 50% (v/v) glycerol. It is supplied at 5-10 units/μL.

DNA polymerase with 5'→3' and 3'→5' exodeoxyribonuclease activities

Performance and quality testing
Endodeoxyribonuclease assay; efficiency of DNase I-dependent nick translation determined.

Unit definition

One unit incorporates 10nMol of total deoxyribonucleotide into acid-precipitable material in 30 min at 37°C using template/primer.

Unit reaction conditions

50mM potassium phosphate (pH 7.0), 6.7mM MgCl2 , 1mM 2-mercaptoethanol, 80μg/mL template/primer, 32μM dTTP, 69nM [3H]dTTP, and enzyme in 100μL for 30 min at 37°C. Also incorporates biotinylated nucleotides.

  • Applications: DNase I-dependent nick translation, second-strand synthesis in cDNA cloning, fill-in of 5' overhangs
  • Source: purified from E. coli expressing the DNA Polymerase I gene on a plasmid

Cloning, Restriction Enzyme Cloning

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Description DNA Polymerase
Form Liquid
Polymerase DNA Polymerase I
Exonuclease Activity 3' - 5', 5' - 3'
Quantity 250 U
Shipping Condition Wet Ice
Hot Start No

For Research Use Only. Not for use in diagnostic procedures.