missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bluo-Gal

Catalog No. 15519028
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15519028 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15519028 Supplier Invitrogen™ Supplier No. 15519028

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bluo-Gal (halogenated indolyl-β-galactoside) is an alternative histochemical substrate for β-galactosidase that produces a darker blue color than X-gal, the more frequently used substrate.

Bluo-Gal (halogenated indolyl-β-galactoside) is an alternative histochemical substrate for β-galactosidase that produces a darker blue color than X-gal, the more frequently used substrate. As with X-gal, Bluo-Gal produces blue (lac+) bacterial colonies and blue (lac+) viral plaques. Bacteria containing active β-galactosidase produce blue colonies when grown on plates containing Bluo-Gal.

Bluo-Gal can also be used as a substrate with the streptavidin-β-galactosidase conjugate in both immunoblot and immunocytochemistry assays. In the presence of β-galactosidase, Bluo-Gal yields a blue precipitate, which is insoluble in alcohols and xylenes. Bluo-Gal is soluble in dimethylformamide.

Primary applications
• Blue-white screening (deeper blue as compared to X-gal)—detect functional β-galactosidase (reporter gene expression)
• Histochemical detection of β-galactosidase activity in fixed tissues
• Bluo-Gal overlay assay

Find the media format that fits your needs
• Ready-to-use media formulations are available, such as LB Broth (Cat. No. 10855001), Terrific Broth (Cat. No. A1374301), or S.O.C recovery medium (Cat. No. 15544034).
• IPTG (Cat. No.15529019) is available for blue-white screening.

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage • Bluo-Gal, substrate for β-galactosidase (1 g)

Store at –5°C to –30°C.

Description Bluo-Gal, Cell-Permeant
Detection Method Colorimetric
Format Bottle
Test Type Blue/White Colony Screening, Immunoblotting, Immunocytochemical Assays
Material Halogenated Indolyl-β-galactoside Substrate
Quantity 1 g
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
How do I resuspend IPTG and X-Gal?

IPTG can be reconstituted in water. Make a stock of 100 mM in water and store working aliquots at -20°C. X-gal can be reconstituted in DMSO, or in a 50:50 mix of DMSO and water. To do the latter, you must dissolve in DMSO first, and then add water to bring up to final volume. It is not necessary to filter sterilize these solutions.

The X-gal solution should be protected from light. To make plates, add 50 ug/ml X-gal and 1 mM (0.24 mg/mL) IPTG to LB/agar that has been cooled down to 50°C. To spread on top of plates, use 50 µl 2% stock of X-gal and 30 µl 100 mM stock of IPTG. 

For Research Use Only. Not for use in diagnostic procedures.