missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Oligo (dT) Primer (50 μM)

These are the same Invitrogen primers that are currently included in the SuperScript IV Kit (Cat. No. 18091050).

Supplier:  Invitrogen™ AM5730G

Encompass

Catalog No. AM5730G


Only null left

Description

Description

These are the same Invitrogen primers that are currently included in the SuperScript IV Kit (Cat. No. 18091050). They are provided at a stock concentration of 50 μM.

Order Info

Shipping Conditions: Dry Ice

Specifications

Specifications

Primer
Dry Ice
Random
Ambion
Store at –20°C.
50 μM
80 μL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.