missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bio-11-UTP (10 mM)

Catalog No. AM8450
Encompass
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8450 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM8450 Supplier Invitrogen™ Supplier No. AM8450

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ambion Bio-11-UTP 10 mM (Biotinylated UTP) is intended for use as substrate in in vitro transcription reactions.

Ambion Bio-11-UTP 10 mM (Biotinylated UTP) is intended for use as substrate in in vitro transcription reactions. These modified nucleotides can be utilized by a variety of RNA polymerases, including T7, T3, and SP6 RNA polymerases, as well as other RNA modifying enzymes. It is provided in one tube containing 25 μL (250 nmol).

Applications
Biotinylated RNA can be used in place of radioactively labeled RNA in many applications, including cRNA amplification and labeling for microarray target preparation. Detection is then carried out via one of a variety of streptavidin-based methods.

Order Info

Shipping Conditions: Dry Ice

Specifications

Product Type Nucleotides
Content And Storage Bio-11-UTP, 10 mM (25 μL)

Store at –70°C.
Shipping Condition Dry Ice
Concentration 10 mM
Label or Dye Biotin
Quantity 25 μL
Product Line Ambion
For Use With (Application) Nucleic Acid Labeling, Oligo Synthesis
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.