missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5-(3-Aminoallyl)-UTP (50 mM)

Catalog No. AM8437
Encompass
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8437 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM8437 Supplier Invitrogen™ Supplier No. AM8437

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ambion modified nucleotides confer unique characteristics to the RNA molecules into which they are incorporated.

Ambion modified nucleotides confer unique characteristics to the RNA molecules into which they are incorporated. Incorporation of an aminoallyl-modified NTP, for example, allows for the subsequent coupling of the reactive amino group to another molecule such as biotin or a Cy dye. Unlike NTPs directly coupled to a dye, aminoallyl-modified NTPs are incorporated almost as efficiently as unmodified NTPs and are much less expensive than dye-coupled NTPs.

Ambion 5-(3-Aminoallyl)-UTP (50 mM) is supplied in a single tube at 50 mM.

Order Info

Shipping Conditions: Dry Ice

Specifications

Product Type Nucleotides
Content And Storage 5-(3-Aminoallyl)-UTP, 50 mM (50 μL)

Store at –70°C.
Shipping Condition Dry Ice
Concentration 50 mM
Label or Dye Aminoallyl
Quantity 50 μL
Product Line Ambion
For Use With (Application) Nucleic Acid Labeling
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.