missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5-(3-Aminoallyl)-dUTP (50 mM)

Catalog No. AM8439
Encompass
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8439 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM8439 Supplier Invitrogen™ Supplier No. AM8439
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

When incorporated into DNA, Ambion 5-(3-aminoallyl)-dUTP provides a reactive group for addition of other chemical groups.

When incorporated into DNA, Ambion 5-(3-aminoallyl)-dUTP provides a reactive group for addition of other chemical groups. The modified dUTP is readily incorporated by reverse transcriptase. The aminoallyl modification enables downstream reaction with amine-reactive compounds such as N-hydroxysuccinimide (NHS) esters; thus aminoallyl-modified cDNA can be labeled with any amine-reactive group moiety.

Use of this modified nucleotide
Use 5-(3-aminoallyl)-dUTP as recommended in your protocol for reverse transcription. Substitution of 30–80% of the dTTP with 5-(3-aminoallyl)-dUTP may be useful, but the exact proportion will depend on experimental goals.

Order Info

Shipping Conditions: Dry Ice

Specifications

Concentration 50 mM
Content And Storage 5-(3-Aminoallyl)-dUTP, 50 mM (50 μL)

Store at –70°C.
Form Liquid
Label or Dye Aminoallyl
Product Line Ambion
Product Type Nucleotides
Purification Method HPLC
Quantity 50 μL
Shipping Condition Dry Ice
For Use With (Application) Nucleic Acid Labeling
Why are there only two amino-modified nucleotides in the dNTP mix for cDNA indirect labeling (aminoallyl-dUTP and aminohexyl-dATP) intead of all four nucleotides being modified? Would more modified nucleotides have an adverse effect on the microarray?

Using four modified nucleotides rather than two can cause fluorescence quenching since the fluorophores are positioned very closely to one another. Use of two amino-modified nucleotides in the cDNA synthesis reaction (rather than one) results in a greater incorporation of fluorescent dye and higher signal intensity with small amounts of RNA starting material. Unbiased incorporation of amino-modified dNTPs and the high efficiency of the coupling reaction result in an even distribution of fluorescent signal and high overall levels of fluorescence, increasing the sensitivity and reproducibility of array hybridizations. In addition to quenching, a very high dye density on the cDNA will interfere with hybridization and, therefore, yield lower microarray fluorescence signals. We found that the use of 2 amino-modified nucleotides (at the appropriate concentration) results in optimal incorporation and high microarray signals.

Can dU-containing DNA produced by amplification with dUTP be cut by restriction enzymes?

You will need to confirm with the manufacturer of the restriction enzyme you are using to find out whether or not it can recognize dUTP-containing DNA.

For Research Use Only. Not for use in diagnostic procedures.