missing translation for 'onlineSavingsMsg'
Learn More

Inulin

Catalog No. AAA1842509
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1842509 10 g
AAA1842518 50 g
AAA1842530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1842509 Supplier Thermo Scientific Chemicals Supplier No. A1842509
Only null left

CAS: 9005-80-5

Mixture of fructose polymers that serves as carbohydrate storage in plantsInulin is used in the treatment of high blood fats, cholesterol and triglycerides. It is also used for weight loss, constipation and also serves as a food additive to improve taste and as a vaccine adjuvant. Further, it is utilized to replace sugar, fat and flour. It is also used to measure kidney function by determining the glomerular filtration rate (GFR). In addition to this, it is used to replace some of the flour in baked goods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mixture of fructose polymers that serves as carbohydrate storage in plantsInulin is used in the treatment of high blood fats, cholesterol and triglycerides. It is also used for weight loss, constipation and also serves as a food additive to improve taste and as a vaccine adjuvant. Further, it is utilized to replace sugar, fat and flour. It is also used to measure kidney function by determining the glomerular filtration rate (GFR). In addition to this, it is used to replace some of the flour in baked goods.

Solubility
Soluble in water.

Notes
Yields D-fructose and D-glucose after acid hydrolysis.Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 9005-80-5
MDL Number MFCD00131407

Specifications

Odor Odorless
Assay Percent Range 0.98
Quantity 10 g
Merck Index 14,5004
Solubility Information Soluble in water.
Chemical Name or Material Inulin
EINECSNumber 232-684-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only