missing translation for 'onlineSavingsMsg'
Learn More

Inulin, MP Biomedicals™

$685.54 - $685.54

Chemical Identifiers

CAS 9005-80-5
Molecular Formula (C6H10O5)n
Synonym Dahlin
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19897180
View Documents
MP Biomedicals Inc
0219897180
100 g
Each for $685.54
Only null left
 
Description

Description

Specifications

Chemical Identifiers

9005-80-5
(C6H10O5)n
Dahlin

Specifications

9005-80-5
(C6H10O5)n
Dahlin
Free-flowing Powder
White
100 g
≈5,000
Inulin
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'