missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Inulin, from chicory root

Catalog No. p-7077658
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457100250 25 g
AC457105000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457100250 Supplier Thermo Scientific Chemicals Supplier No. 457100250
Only null left

CAS: 9005-80-5

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9005-80-5

Specifications

Loss on Drying 10% max. (2 g, 105°C, 2 hrs)
Infrared Spectrum Authentic
Quantity 25 g
Chemical Name or Material Inulin
EINECSNumber 232-684-3

RUO – Research Use Only