missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Introductory Molecular Model Set - 31 atoms

Catalog No. S41610
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S41610 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S41610 Supplier Eisco™ Supplier No. SET00615

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

12 White 1 Face Hydrogen (30mm), 4 Black 4 Face Carbon (38 mm), 1 Black 2 Face Carbon (38 mm), 2 Red 2 Face Oxygen (38 mm), 7 Red 1 Face Oxygen (38 mm), 1 Blue 3 Face Nitrogen (38 mm), 2 Blue 1 Face Nitrogen (38 mm), 2 Green 1 Face Chlorine (50 mm), 20 Short Links (16 mm)

Introductory molecular model set designed for introductory study of the geometry of molecules (VESPR). Includes 31 atoms and 20 short links. Build multiple models of simple molecules in their compact representation, present molecules acting as reagents or products of simple chemical reactions

  • Molecular model set includes 31 atoms, 20 short links
  • Supplied in 5 compartment plastic storage case
  • Designed for introductory study of the geometry of molecules (VESPR)
  • Build multiple models of simple molecules in their compact representation
  • Present molecules acting as reagents or products of simple chemical reactions
  • Achieve a molecular representation of pure or polluted air
  • Contents: 12 White 1 Face Hydrogen (30mm), 4 Black 4 Face Carbon (38 mm), 1 Black 2 Face Carbon (38 mm), 2 Red 2 Face Oxygen (38 mm), 7 Red 1 Face Oxygen (38 mm), 1 Blue 3 Face Nitrogen (38 mm), 2 Blue 1 Face Nitrogen (38 mm), 2 Green 1 Face Chlorine (50 mm), 20 Short Links (16 mm).

Warranty

1 year limited manufacturer warranty

Specifications

Color Gray, White, Green, Blue, Red, Black
For Use With (Application) Building molecular models
Height (Metric) 5.72 cm
Includes 12 White 1 Face Hydrogen (30mm), 4 Black 4 Face Carbon (38 mm), 1 Black 2 Face Carbon (38 mm), 2 Red 2 Face Oxygen (38 mm), 7 Red 1 Face Oxygen (38 mm), 1 Blue 3 Face Nitrogen (38 mm), 2 Blue 1 Face Nitrogen (38 mm), 2 Green 1 Face Chlorine (50 mm), 20 Short Links (16 mm)
Length (Metric) 23.2 cm
Material Plastic
Quantity 1 Ea.
Size 31 Atoms
Weight (Metric) 740 g
Width (Metric) 16.84 cm
Product Type Molecular Model Set
Show More Show Less