Learn More
Introductory Earth Science Rock and Mineral Collection
Catalog No. S45051
Change view
Click to view available options
For Use With (Application):
Earth Science-Rock and Minerals
Catalog No. For Use With (Application)
S45051 Earth Science-Rock and Minerals
1 options
Catalog No. S45051 Supplier Geoscience Industries Supplier No. S45051
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
A handbook, streak plate, magnifier, and magnet come with the collection.
Designed to match most introductory-level textbooks
This all-in-one collection covers a broad range of demonstration needs! The 75 specimens for observation, comparison, and testing include 13 igneous rocks; 14 sedimentary rocks; 9 metamorphic rocks; 17 rock-forming minerals; 9 samples from the hardness scale (diamond omitted); and 13 examples of important physical properties (specific gravity, tenacity, cleavage, fracture, luster, streak, refraction, magnetism, and acid test).
Specifications
For Use With (Application) Earth Science-Rock and Minerals
Includes A handbook; streak plate; magnifier; and magnet.