Learn More
International Enviroguard™ MicroGuard CE™ Hoods
Catalog No. 19472564
Change view
Click to view available options
Packaging Type:
Individually Packaged
Catalog No. Packaging Type
19472564 Individually Packaged
1 options
Catalog No. 19472564 Supplier International Enviroguard™ Supplier No. CE8063CI
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MicroGuard CE™ Hoods, disposable protective clothing designed with your comfort in mind. Made from high MVTR, innovative fabric.
  • Microporous, clean processed
  • Fabric keeps dust and liquids out while allowing air to flow freely
  • Ideal for non-hazardous work environments
  • Provide reliable liquid and particulate protection
  • Tunnelized elastic offer secure closure and stronger seams
  • Bulk or individually packaged to help prevent cross-contamination
  • Environmentally friendly, can be recycled after use
Specifications
Product Type Open Face
Size Universal
Material Microporous
Color White
Closure Type Elastic
Model CE8063CI
Seam Type Serged
Packaging Type Individually Packaged
Quantity 100/Cs.
Includes Low linting, liquid barrier, tunnelized elastic face
Hood Type Clean Processed
Show More Show Less