Learn More
Interlocking Cube Grams
Catalog No. S94318
Change view
Click to view available options
Quantity:
100/Pk.
200/Pk.
500/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S94318 500/Pk.
S94316 100/Pk.
S94317 200/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog No. S94318 Supplier Customer Channels Group (Ccg) Supplier No. 48179
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Each cube measures one cubic centimeter and weighs one gram.
Ten brightly colored cubes have receptacles and fittings to easily chain together.
Demonstrate the following concepts:
  • Determining conservation of mass
  • Factoring
  • Squaring fractions
  • Pythagorean Theorem
  • Commutative and associative properties
Specifications
Quantity 500/Pk.
For Use With (Application) To demonstrate determining conservation of mass, factoring, squaring fractions, Pythagorean theorem, commutative and associative properties
Product Type Interlocking Centimeter Cube
Includes Ten brightly colored cubes with receptacles and fittings
Weight (English) 0.002 lb.
Weight (Metric) 0.9 g