Learn More
BUCHI Rotavapor I-300 Vacuum Controller and Interface
Catalog No. 05000003
Change view
Click to view available options
Includes:
Kit for V-300/600 Woulff Bottle
Standalone
Valve Unit
For Use With (Equipment):
R-300
R-300 with Non-BUCHI Vacuum Pumps
R-300, V-300, V-600
Model:
Pro
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes For Use With (Equipment) Model
05000003 Standalone R-300, V-300, V-600 Pro
05000006 Valve Unit R-300 with Non-BUCHI Vacuum Pumps Pro
10987135 Kit for V-300/600 Woulff Bottle R-300 Pro
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05000003 Supplier BUCHI Supplier No. 11I300P00
Only null left
The BUCHI Interface I-300 controls all distillation parameters of a Rotavapor R-300 system including the optional V-300 Vacuum Pump and F-3XX series chillers.
  • Easy plug and play integration into a Rotavapor R-300 system at any time
  • Central control of all process parameters such as rotation speed, temperature (heating, cooling, vapor) and pressure
  • Remote monitoring on smart phones including push notifications and live view
  • Modes: manual, timer, continuous
  • Flexibility of the interface allows easy installation and convenient operation from multiple positions:
    • Rotavapor handle
    • Vacuum pump
    • Fume hood or lab bench wall
  • Stand-alone operation to locally control a central vacuum system
  • Synchronized processing thanks to centrally controlled rotary evaporator, heating bath, speed-controlled vacuum pump and recirculating chiller
  • Unattended operation (optional):
    • Automatic distillation with CloudDest sensor
    • Foam sensor for foaming samples
Specifications
Material Plastic
Type Vacuum Controller/Interface
For Use With (Equipment) R-300, V-300, V-600
Includes Standalone
Model Pro