missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ Interest

Catalog No. 17000046
Change view
Click to view available options
Volume (Metric):
208 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
17000046 208 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17000046 Supplier Diversey™ Supplier No. 57765010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Interest detergent is a low foam liquid chlorinated cleaner designed for clean-in-place (CIP), spray washing and mechanical cleaning of dairy, beverage, meat and food processing equipment applications across the food and beverage industries.

  • Heavy duty CIP cleaner for processing
  • Hard water tolerant, ensuring scale free equipment and improved operational efficiency
  • Low foaming under high turbulence making it easier to rinse and improving cleaning efficiency
  • Hard water tolerant, ensuring scale-free equipment and improved operational efficiency

Specifications

Product Type Chlorinated Alkaline
pH >11
Sterility Non-sterile
Appearance Yellow
Certifications/Compliance CFIA, Kosher, NSF, Halal
Container Type Barrel
Content And Storage Store in corrosive-resistant container with a resistant inner liner.
For Use With (Application) CIP Process
Quantity 1 Ea.
Volume (English) 55 gal.
Volume (Metric) 208 L
Show More Show Less