missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INTELLIFLEX Performance Verification Kit

Catalog No. IFXPVERK20
Change view
Click to view available options
Quantity:
20 reactions kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IFXPVERK20 20 reactions kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IFXPVERK20 Supplier Invitrogen™ Supplier No. IFXPVERK20

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The INTELLIFLEX Performance Verification Kit is for use with the Luminex xMAP INTELLIFLEX and Luminex xMAP INTELLIFLEX DR-SE systems and is used to verify calibration of these instruments.

The INTELLIFLEX Performance Verification Kit is for use with the Luminex xMAP INTELLIFLEX and Luminex xMAP INTELLIFLEX DR-SE systems and is used to verify calibration of these instruments. The Performance Verification Kit contains all reagents needed for verifying the calibration of these INTELLIFLEX systems.

Calibration is important to ensure that the optical system functions effectively and that different INTELLIFLEX systems report similar results. The Performance Verification Kit helps ensure that the classification channels (CL1, CL2, and CL3), doublet discriminator channel (DD), and reporter channels (RP1 and RP2) are all performing as intended. The kit includes reagents to verify the calibration and optical integrity of the INTELLIFLEX system, as well as reagents to verify the fluidics channels using observations of pressure, flow rate, and carryover from well to well. The verification reagents consist of mixtures of different microspheres internally labeled with classification and reporter dyes. The classification control microspheres verify the integrity of the classification and doublet discriminator channel, as well as classification efficiency and misclassification. The reporter verifier microspheres verify the integrity of the reporter channels. The fluidics microspheres verify the integrity of the system fluidics including well to well carryover.

Please refer to the INTELLIFLEX Performance Verification Kit package insert for proper use of the INTELLIFLEX Performance Verification Kit.

Specifications

For Use With (Equipment) Luminex™ xMAP™ INTELLIFLEX, Luminex™ xMAP™ INTELLIFLEX DR-SE
No. of Tests 20 Calibrations
Quantity 20 reactions kit
Content And Storage • xMAP INTELLIFLEX Verifier, 10 mL
• xMAP Fluidics 1 Microspheres, 5 mL
• xMAP Fluidics 2 Microspheres, 5 mL
• 25 Strips wells

Each kit will provide 20 calibrations.
Store in the dark at 2°C to 8°C.
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.