missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INTELLIFLEX Calibration Kit

Catalog No. IFXCALK20
Change view
Click to view available options
Quantity:
20 reactions kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IFXCALK20 20 reactions kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IFXCALK20 Supplier Invitrogen™ Supplier No. IFXCALK20

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The INTELLIFLEX Calibration Kit is for use with the Luminex xMAP INTELLIFLEX and Luminex xMAP INTELLIFLEX DR-SE systems and is used to calibrate the optics of these systems.

The INTELLIFLEX Calibration Kit is for use with the Luminex xMAP INTELLIFLEX and Luminex xMAP INTELLIFLEX DR-SE systems and is used to calibrate the optics of these systems. The calibration kit contains all reagents needed for calibration of these systems.

Calibration is important to ensure that the optical system functions effectively and that different INTELLIFLEX systems report similar results. Calibrating the INTELLIFLEX System normalizes the settings for classification channels (CL1, CL2, and CL3), the doublet discriminator channel (DD), and the reporter channels (RP1 and RP2).

Please refer to the INTELLIFLEX Calibration Kit package insert for proper use of the INTELLIFLEX Calibration Kit.

Specifications

For Use With (Equipment) Luminex™ xMAP™ INTELLIFLEX, Luminex™ xMAP™ INTELLIFLEX DR-SE
No. of Tests 20 Calibrations
Quantity 20 reactions kit
Content And Storage • xMAP INTELLIFLEX Calibrator, 10 mL
• 25 Strips wells

Each kit will provide 20 calibrations.
Store in the dark at 2°C to 8°C.
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.