Learn More
ACROBiosystems Inc Recombinant Protein;Human Integrin alpha 8 beta 1 (ITGA8&ITGB1) Heterodimer Protein;HEK293;1MG;IT1,Integrin alpha 8 beta 1,ITGA8&ITGB1,ITGA8&B1
Catalog No. 501658150
Encompass
Recombinant Protein;Human Integrin alpha 8 beta 1 (ITGA8&ITGB1) Heterodimer Protein;1MG;Human ITGA8&ITGB1 Heterodimer Protein (IT1-H52W9) is expressed from human 293 cells (HEK293). It contains AA Phe 39 - Leu 1012 (ITGA8) & Gln 21 - Asp 728 (ITGB1) (Accession # P53708-1 (ITGA8) & P05556-1 (ITGB1)).;Human ITGA8 & ITGB1 Heterodimer Protein, produced by co-expression of ITGA8 and ITGB1, has a calculated MW of 114.6 kDa (ITGA8) & 83.7 kDa (ITGB1). Subunit ITGA8 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 115-155 kDa due to glycosylation.
Catalog No. 501658150 Supplier ACROBiosystems Inc Supplier No. IT1H52W91MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Human Integrin alpha 8 beta 1 (ITGA8&ITGB1) Heterodimer Protein;1MG;Human ITGA8&ITGB1 Heterodimer Protein (IT1-H52W9) is expressed from human 293 cells (HEK293). It contains AA Phe 39 - Leu 1012 (ITGA8) & Gln 21 - Asp 728 (ITGB1) (Accession # P53708-1 (ITGA8) & P05556-1 (ITGB1)).;Human ITGA8 & ITGB1 Heterodimer Protein, produced by co-expression of ITGA8 and ITGB1, has a calculated MW of 114.6 kDa (ITGA8) & 83.7 kDa (ITGB1). Subunit ITGA8 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 115-155 kDa due to glycosylation.