Recombinant Protein;Human Integrin alpha 5 beta 1 (ITGA5&ITGB1) Heterodimer Protein;100UG;Recombinant Human ITGA5 & ITGB1 Heterodimer Protein (IT1-H52W5) is expressed from human 293 cells (HEK293). It contains AA Phe 42 - Tyr 995 (ITGA5) & Gln 21 - Asp 728 (ITGB1) (Accession # P08648-1 (ITGA5) & P05556-1 (ITGB1)).;Human ITGA5 & ITGB1 Heterodimer Protein, produced by co-expression of ITGA5 and ITGB1, has a calculated MW of 104.1 kDa (ITGA5) & 83.7 kDa (ITGB1). Subunit ITGA5 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 117-130KDa (ITGA5) and 100-116 kDa (ITGB1) respectively due to glycosylation.;Immobilized fibronectin at 10 g/mL (100 L/well) can bind Human ITGA5 & ITGB1 (Cat. No. IT1-H52W5) with a linear range of 0.01-0.156 ug/mL (QC tested).