missing translation for 'onlineSavingsMsg'
Learn More

Integra™ Miltex™ Surgical Instrument Cleaner

Catalog No. p-4184804
Change view
Click to view available options
Volume (English):
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
12460424 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12460424 Supplier Integra™ Miltex™ Supplier No. MLT3725
Only null left

Neutral pH (7) detergent

  • To remove organic material from instruments
  • Biodegradable
  • Rinses-free, no residue
  • Concentrated
  • Phosphate-free

Specifications

Form Liquid
pH 7
Container Type Pump bottle
For Use With (Application) To remove organic material from instruments
Volume (English) 1 gal.