Learn More
Integra™ Miltex™ Standard Biopsy Punches
Stainless-steel razor sharp cutting edge
$450.41 - $652.76
Specifications
Blade Material Stainless Steel
Product Type Biopsy Punch
For Use With (Application) Offer the Convenience of Single-use Disposability without Sacrificing quality, Feel or Performance
Handle Type Ribbed
Description Seamless
View More Specs
Products 11
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
12460399
View Documents Promotion Details
Integra™
MLT3331
0.2 cm
Pack of 50 Each for $517.50
Only null left
12460400
View Documents Promotion Details
Integra™
3331A
0.15 cm
Pack of 50 Each for $652.76
Only null left
12460401
View Documents Promotion Details
Integra™
3331AA
0.1 cm
Pack of 50 Each for $450.41
Only null left
12460404
View Documents Promotion Details
Integra™
3331B
0.25 cm
Pack of 50 Each for $450.41
Only null left
12460406
View Documents Promotion Details
Integra™
3332
0.3 cm
Pack of 50 Each for $517.50
Only null left
12460408
View Documents Promotion Details
Integra™
MLT3333
0.35 cm
Pack of 50 Each for $450.41
Only null left
12460409
View Documents Promotion Details
Integra™
MLT3334
0.4 cm
Pack of 50 Each for $517.50
Only null left
12460414
View Documents Promotion Details
Integra™
MLT3338
0.2, 0.3, 0.4, 0.5, 0.6 cm
Pack of 50 Each for $517.50
Only null left
12460412
View Documents Promotion Details
Integra™
MLT3336
0.6 cm
Pack of 50 Each for $517.50
Only null left
12460411
View Documents Promotion Details
Integra™
MLT3335
0.5 cm
Pack of 50 Each for $550.38
Only null left
12460413
View Documents Promotion Details
Integra™
MLT3337
0.8 cm
Pack of 50 Each for $517.50
Only null left
Description
Description
Stainless-steel razor sharp cutting edge
  • Sealed in a sterile, peel-apart, see-through package
  • Sterilized by ethylene oxide
  • Single use
Order Info
Each box contains 50 sterile color-coded blister packs
Specifications
Specifications
Stainless Steel
Offer the Convenience of Single-use Disposability without Sacrificing quality, Feel or Performance
Seamless
Sterile
Biopsy Punch
Ribbed
Single-use
50/Pk.
Videos
Safety and Handling
Safety and Handling
ShelfLife : 5 Years