Learn More
Integra™ Miltex™ O'Brien Stitch Scissors
Angled, sharp points
Supplier:  Integra™ Miltex™ MLT9110
Catalog No. 12-460-578
Only null left
Explore available promotions
Description
Description
  • High-quality surgical-grade stainless steel construction provides optimal performance
  • Fully tempered precision screw locks maintain perfect alignment
  • Ergonomic design combines weight, balance and feel for optimal comfort and performance
  • Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
  • Length: 3.75 in. (9.5cm)
Specifications
Specifications
Straight
9.5 cm
Sharp/Sharp
3.75 in.
Surgical, Stitch
Product Suggestions
Product Suggestions