missing translation for 'onlineSavingsMsg'
Learn More

Integra™ Miltex™ Magnifying Loupes

Quickly and easily locked into personal setting using a unique locking knob

Supplier:  Integra™ Miltex™ 15014

 View more versions of this product

Catalog No. 12460198


Only null left

Description

Description

  • Dual hinge design
  • Locking interpupillary knob
  • Double locking convergence angle
Specifications

Chemical Identifiers

Loupe, Magnifying
For endodontic procedures

Specifications

40.64 to 50.8 cm
Loupe, Magnifying
4.6 in.
For endodontic procedures
16 to 20 in.
11.68 cm
X-Large
Black
2.5x
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions