missing translation for 'onlineSavingsMsg'
Learn More

Integra™ Miltex™ Iris Scissors

All-purpose scissors

$195.12 - $534.29

Specifications

Grade Premium
For Use With (Application) Surgical, ophthalmic
View More Specs

Products 5
Catalog Number Mfr. No. Length (Metric) Blade Style Material Product Type Price Quantity  
Catalog Number Mfr. No. Length (Metric) Blade Style Material Product Type Price Quantity  
12-460-104
View Documents
Integra™ Miltex™
5304
11.43 cm Straight Stainless Steel Iris Scissors
Each for $263.76
Only null left
 
12-460-270
View Documents
Integra™ Miltex™
181398
8.8 cm Curved Stainless Steel Iris Scissors
Each for $384.47
Only null left
 
12-460-490
View Documents
Integra™ Miltex™
5300
10.16 cm Straight Stainless Steel Iris Scissors
Each for $231.06
Only null left
 
12-460-493
View Documents
Integra™ Miltex™
5306
11.43 cm Curved Stainless Steel Iris Scissors
Each for $195.12
Only null left
 
12-460-514
View Documents
Integra™ Miltex™
5SC304
11.43 cm Straight Stainless Steel SuperCut Iris Scissors
Each for $534.29
Only null left
 
Description

Description

  • High-quality surgical-grade stainless steel construction provides optimal performance
  • Fully tempered precision screw locks maintain perfect alignment
  • Ergonomic design combines weight, balance and feel for optimal comfort and performance
  • Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
  • Straight or curved patterns
  • Tungsten carbide inserts seamlessly bonded to upper and lower blades and feature gold ring handles
  • SuperCut scissors have a specially designed razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to prevent tissue slippage

Order Info

Cold or chemical sterilization not recommended for tungsten carbide scissors. SuperCut scissiors designed to cut tissue only; not recommended for use on suture material. Dropping or mishandling will cause the razor-shapr edge to nick or become irregular.

Specifications

Specifications

Premium
Surgical, ophthalmic
Videos
SDS
Documents

Documents