missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integra™ Miltex™ Baumgartner Needle Holder
Specifications
Specifications
| Product Type | Baumgartner Needle Holder |
| Material | Stainless Steel |
| Style | Serrated |
| Length (English) | 5.5 in. |
| Length (Metric) | 14 cm |