missing translation for 'onlineSavingsMsg'
Learn More

Integra™ Miltex™ Alm Retractor

Separates and holds tissue and organs

$897.94 - $1,051.59

Specifications

Product Type Alm Retractor
Grade Premium
Material Stainless Steel
For Use With (Application) Separates and holds tissue and organs
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
12460180
View Documents Promotion Details
Integra™ Miltex™
MLT1112
2.75 in.
Each for $897.94
Only null left
 
12460181
View Documents Promotion Details
Integra™ Miltex™
MLT1113
3.87 in.
Each for $1,051.59
Only null left
 
Description

Description

  • Self-retaining
  • High-quality surgical-grade stainless steel
Specifications

Specifications

Alm Retractor
Premium
Stainless Steel
Separates and holds tissue and organs
Videos
SDS
Documents

Documents