missing translation for 'onlineSavingsMsg'
Learn More

Sol-Guard™ Insulin Safety Syringe with Fixed Hypodermic Needle

Catalog No. 22024442
Change view
Click to view available options
Needle Gauge:
29 G
Volume (Metric):
0.3 mL
0.5 mL
1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Volume (Metric)
22024442 29 G 0.3 mL
22024443 29 G 0.5 mL
22024445 29 G 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22024442 Supplier Sol-Guard™ Supplier No. 200003SG
Only null left

Desinged to provide accurate and comfortable insulin administration. Sol-Guard Safety Insulin Syringes have features which help to prevent needlestick injuries.

  • Needles are tri-bevel and equipped with plunger lock and break feature that prevents reuse of syringe
  • Low dead space - improved accuracy and less medication waste when compared to conventional syringes
  • Bold and clear graduated insulin unit markings
  • Safety shield feature that slides over the needle to prevent needlestick injuries before and after medication delivery

Specifications

Autoclavable Not Autoclavable
Material Polypropylene
Needle Gauge 29 G
Latex Free Yes
Disposable Single-use
Volume (Metric) 0.3 mL
Length (Metric) Needle 12.7 mm
Needle Point Style Triple-bevel
For Use With (Equipment) Insulin
Connection Type Fixed
Graduated Yes
Sterility Sterile
Show More Show Less