Good for instruments up to 16cm long, this plastic case is safe, secure, and slotted for placement of individual surgical instruments aiding in the prevention of tip damage. Case is autoclavable but should not be used for sterilizing instruments.
Catalog No.
NC1832124
Supplier
Fine Science ToolsSupplier No.
2083006
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Good for instruments up to 16cm long, this plastic case is safe, secure, and slotted for placement of individual surgical instruments aiding in the prevention of tip damage. Case is autoclavable but should not be used for sterilizing instruments.