missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ InstantOne ELISA Wash Buffer (10X)
Description
- If washing wells with a microplate washer, use 3x washes with a 10-second mixing cycle
- Included with every InstantOne ELISA sold. Offered as a stand-alone item for cases where experimental procedures and/or automation require much higher volumes of wash buffer
Specifications
Specifications
| Chemical Name or Material | Wash Buffer |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | InstantOne ELISA Cell Signaling assays |
| Physical Form | Liquid |
| Quantity | 500 mL |
For Research Use Only. Not for use in diagnostic procedures.