Learn More
Pentair Aquatic Instant Ocean Sea Salt, 50 Gallon
Catalog No. NC0985949
Encompass
nstant Ocean has been the preferred salt mix of public aquariums and home aquarists for many years. It is scientifically blended to contain every minor and major trace element required to make artificial seawater. Instant Ocean salt contains no phosphates or nitrates, chemicals that can promote the growth of algae and damage coral. This salt also dissolves quickly without leaving insoluble residue. It contains no chlorine remover.
Catalog No. NC0985949 Supplier Pentair Aquatic Supplier No. IS50
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
nstant Ocean has been the preferred salt mix of public aquariums and home aquarists for many years. It is scientifically blended to contain every minor and major trace element required to make artificial seawater. Instant Ocean salt contains no phosphates or nitrates, chemicals that can promote the growth of algae and damage coral. This salt also dissolves quickly without leaving insoluble residue. It contains no chlorine remover.