missing translation for 'onlineSavingsMsg'
Learn More

Interscience InstaBAG BPW

Catalog No. 16949063
Change view
Click to view available options
Capacity (English):
3 oz.
7.6 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
16949063 3 oz.
16949061 7.6 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16949063 Supplier Interscience Supplier No. 114090

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The InstaBAG BPW is a filter bag with dehydrated buffered peptone water.

The InstaBAG BPW is a filter bag that includes a dehydrated pre-dosed media, allowing a 2-in-1 process: media and sample preparation for microbiological analyses.

  • Filter bag with dehydrated pre-dosed media
  • Type of dehydrated media: buffered peptone water
  • Filter porosity: <250 microns
  • Compatible with any laboratory blender

Available as 90 mL bag or 225 mL bag.

Specifications

Autoclavable Not Autoclavable
Color Clear
Disposable Single-use
Sterility Sterile
Latex Free Yes
Thickness (English) 3 mil
Length (English) 12 in.
Width (English) 7.48 in.
Product Type Blender Bag
Capacity (English) 3 oz.
Capacity (Metric) 90 mL
Length (Metric) 300 mm
Width (Metric) 190 mm
Thickness (Metric) 72 μm
Show More Show Less