LaMotte™ Insta-Test™ Water Test Strips
Perform safe, quick and easy water quality checks with test strips.
$33.51 - $42.21
Catalog Number | Mfr. No. | For Use With (Application) | Range | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Range | Price | Quantity | ||||
S66231
|
LaMotte™
EMD2974 |
Drinking, food/beverage, pool water testing | 4 to 10 (4, 5, 6, 7, 8, 9, 10pH) |
Each for $40.23
|
|
S66234
|
LaMotte™
EPD2987G |
Drinking, food/beverage, pool water testing | 4 to 10 (pH), 0 to 50 (TCl) |
Each for $33.51
|
|
S66235
|
LaMotte™
2996 |
Drinking water testing | 0 to 50 (Nitrate), 0 to 10 (Nitrite) |
Each for $42.21
|
|
Description
- Monitor water without having to use reagents or field kits
Order Info
Each vial of test strips contains 50 tests per factor, except S66233, which contains 25 tests per factor.
Specifications
Water Test Strip |