missing translation for 'onlineSavingsMsg'
Learn More

LaMotte™ Insta-Test™ Water Test Strips

Catalog No. S66235
Change view
Click to view available options
Analytes:
Hardness
Nitrate, Nitrite
pH
Total Chlorine, pH
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Analytes
S66235 Nitrate, Nitrite
S66231 pH
S66230 Hardness
S66234 Total Chlorine, pH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S66235 Supplier LaMotte™ Supplier No. 2996
Only null left

Perform safe, quick and easy water quality checks with Insta-Test™ Water Test Strips.

  • Monitor water without having to use reagents or field kits.

Order Info

Each vial of test strips contains 50 tests per factor, except S66233, which contains 25 tests per factor.

Specifications

Analytes Nitrate, Nitrite
Format Strip
For Use With (Application) Drinking water testing
Quantity 50 Pk.
Range 0 to 50 (Nitrate), 0 to 10 (Nitrite)
Type Water Test Strip