missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LaMotte™ Insta-Test™ Water Test Strips
Description
- Monitor water without having to use reagents or field kits.
Order Info
Each vial of test strips contains 50 tests per factor, except S66233, which contains 25 tests per factor.
Specifications
Specifications
| Analytes | pH |
| Format | Strip |
| For Use With (Application) | Drinking, food/beverage, pool water testing |
| Quantity | 50 Pk. |
| Range | 4 to 10 (4, 5, 6, 7, 8, 9, 10 pH) |
| Type | Water Test Strip |