missing translation for 'onlineSavingsMsg'
Learn More

LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips

Catalog No. S72328
Change view
Click to view available options
Parameter Specifications:
Chlorine - free, low range
Quantity:
25 Strips/Vial
Range:
0-0.5-1-3-5-10 ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Parameter Specifications Quantity Range
S72328 Chlorine - free, low range 25 Strips/Vial 0-0.5-1-3-5-10 ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72328 Supplier LaMotte™ Supplier No. 2964G/EMD
Only null left

For water quality testing.

  • No reagents or field kits required for most strips
  • Chemically treated pads on plastic strips produce color change
  • Pop-top vials have molded desiccant liner and hinged snap cap to protect from moisture
  • As little as 10-second development time
  • Test strips for testing water: wastewater, industrial water boiling and cooling, drinking water, laundry and sanitation †

Specifications

For Use With (Application) Water quality testing
Parameter Specifications Chlorine - free, low range
Quantity 25 Strips/Vial
Range 0-0.5-1-3-5-10 ppm
Storage Requirements Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
Type Single Factor Test Strip