Learn More
LaMotte™ Insta-Test™ Chlorine Test Strips
Test for free and total chlorine with one strip.
Supplier: LaMotte™ 3027GEMD
Description
- Range and sensitivity is 0, 0.5, 1, 3, 5, 10 ppm.
Specifications
| Strips | |
| 25 | |
| 25 Pk. | |
| 0, 0.5, 1, 3, 5, 10 ppm |