Natural Arkansas oil stones feature unequalled fineness, hardness and structural density. When used with instrument oil, they will produce the sharpest cutting edge. Triangle, 6mmx9cm.
Catalog No.
NC9487121
Supplier
Fine Science ToolsSupplier No.
2900812
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Natural Arkansas oil stones feature unequalled fineness, hardness and structural density. When used with instrument oil, they will produce the sharpest cutting edge. Triangle, 6mmx9cm.