Learn More
Thermo Scientific™ InSight Sensors
Catalog No. 11676399
Change view
Click to view available options
Description:
CT Smart Cable (spare part)
RTD Smart Cable (spare part)
Temp sensor PT100 (spare part)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
11676399 RTD Smart Cable (spare part)
11676402 CT Smart Cable (spare part)
11676398 Temp sensor PT100 (spare part)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11676399 Supplier Thermo Scientific™ Supplier No. INSPTSMRT001
Thermo Scientific™ InSight sensors work with the mote transmitters, relaying data to the InSight Cloud service.
Thermo Scientific™ InSight sensors relay data from the motes on your wireless account to the InSight Cloud service. In the event of internet failure, Insight Cloud will notify IT and the InSight link will log sensor data for data backfill on recovery.
InSight Analytics:
InSight Analytics, powered by KLATU™, is a system designed to collect, analyze and make efficiency and operating cost recommendations from multiple asset inputs and events such as:
  • Equipment critical parameters
  • Electrical use and energy consumption
  • Asset utilization
  • Environmental conditions
InSight Analytics monitors your laboratory equipment 24 hours a day, seven days a week and alerts you when the thresholds are exceeded. The data from InSight Analytics is run through a number of algorithms utilizing the InSight software. With InSight Analytics, you can determine and compare the assets' performance against industry performance standards.
How InSight Analytics works:
  • Data is sent wirelessly from the InSight mote via your Wi-Fi network to the InSight web platform fpr storage.
  • Once in the Cloud, the data is analyzed by the system, providing real-time comparison of the laboratory equipment's health to industry averages.
  • This data analysis enables you to make knowledge-based asset decisions based on key performance indicators such as product service and support, service vs. buy decisions and remote system health monitoring and trouble-shooting for service providers.
Specifications
Description RTD Smart Cable (spare part)
For Use With (Application) Wireless Monitoring, Asset Management Solution
Not available outside North America