missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ INSIG-1 Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 3638 |
| Product Line | Novus |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB110-55244 |
| Gene Alias | CL6, CL-6, INSIG-1, INSIG-1 membrane protein, insulin induced gene 1, insulin-induced gene 1 protein, MGC1405 |
| Gene Symbol | INSIG1 |
| Quantity | 0.1 mg |
| Research Category | Cholesterol Metabolism, ER Markers, Lipid and Metabolism, Signal Transduction |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |