Learn More
Chemglass Life Sciences Glass Insert, 0.35mL, Flat Bottom DFS Item
Supplier:  Chemglass Inc CV14340630
Glass Insert, 0.35mL, Flat Bottom. Both glass and plastic inserts have been engineered to fit the large opening in vials containing a nominal I.D. of 6mm. Inserts are available assembled with a polymer bottom spring or patented Top Spring™. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.
Catalog No. 50-230-855
Only null left
Explore available promotions