missing translation for 'onlineSavingsMsg'
Learn More

Ritmed™ Insemination Gloves

Catalog No. 296369821
Change view
Click to view available options
Color:
Red
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
296369821 Red
296369820 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 296369821 Supplier Ritmed™ Supplier No. 9965

Designed to provide full arm coverage up to the shoulder for veterinary insemination. Also suitable for general use in farming, agriculture, nursing homes, and residential care settings.

  • Versatile glove suitable for use in agriculture, veterinary, nursing homes, and residential care settings.
  • Extended-length design provides full arm coverage for insemination procedures.
  • Ambidextrous glove with a seamless cuff that extends into a protective sleeve.
  • Latex-free and powder-free to minimize the risk of allergic reactions.

Specifications

Chlorinated No
Accelerator Free No
Applications Laboratory
AQL None
Color Red
Disposable Single-use
Exterior Finish Smooth
Latex Free Yes
Material Poly-Vinyl
Powdered Powder Free
Product Type Insemination Gloves
Size Universal
FDA 510 (k) No
Length (English) 36.2 in.
Length (Metric) 92 cm
Show More Show Less