missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Insect Pins with Yellow Epoxy Head
Description
- Steel pins
- Yellow epoxy heads
- Available in sizes 000 through 5
Specifications
Specifications
| Product Type | Insect Pins |
| Autoclavable | Not Autoclavable |
| Coating | Black Corrosion Resistant |
| Color | Silver |
| Material | Steel |
| For Use With (Application) | Dissection |
| Includes | Yellow epoxy head |
| Size | 000 |