Learn More
LaMotte™ Aquatic Macroinvertebrate Life Cycle and Habitat Flashcards
Catalog No. S67780
Change view
Click to view available options
Class Size:
1 Class
Catalog No. Class Size
S67780 1 Class
1 options
Catalog No. S67780 Supplier LaMotte™ Supplier No. 5946
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Discover the world of the macroinvertebrate.
  • Insect Life Cycle and Habitat Flashcards:
  • 22 full-color laminated flashcards
  • Help to illustrate the life cycle and habitats of freshwater aquatic macroinvertebrates
  • Flashcards feature photos of both immature and adult insect stages, detailed life cycle and habitat information
  • Manual contains:
  • Facts on insect characteristics, life cycles, taxonomy, classification, food webs and how macroinvertebrates can indicate water quality; instructions for flashcard activities
  • Can be used as an extension to the Freshwater Aquatic Macroinvertebrate: Identification Flashcards and the Leaf Pack Experiments Stream Ecology Kit
Insect Identification Flashcards:
  • 18 full-color laminated flashcards
  • Can be used alone or as an addition to any macroinvertebrate activity
  • Flashcards feature: a full-color photograph of a common macroinvertebrate in its natural environment on front and a list of dichotomous characteristics, a measuring scale and detailed drawing on the back
  • Included in the Leaf Pack Experiments Stream Ecology Kit
  • Includes helpful notes that prevent confusion between macroinvertebrates with a similar appearance
Specifications
Includes Insect Life Cycle and Habitat Flashcard Kit
Class Size 1 Class
Product Type Insect Life Cycle and Habitat Flashcard Kit