missing translation for 'onlineSavingsMsg'
Learn More

Inosine 5'-Triphosphate Trisodium Salt Hydrate 85.0+%, TCI America™

Catalog No. I0038100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0038100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0038100MG Supplier TCI America Supplier No. I0038100MG
Only null left

Chemical Identifiers

CAS 35908-31-7
Molecular Formula C10H15N4NaO14P3
Molecular Weight (g/mol) 531.16
MDL Number MFCD00084678,MFCD00084678
InChI Key AUOLCGMICMHAHB-UHFFFAOYNA-N
Synonym inosine-5-triphosphate, trisodium salt
PubChem CID 133109127
IUPAC Name sodium ({[({[3,4-dihydroxy-5-(6-oxo-6,9-dihydro-3H-purin-9-yl)oxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)phosphonic acid
SMILES [Na+].OC1C(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)OC(C1O)N1C=NC2=C1NC=NC2=O

Specifications

Color White
Quantity 100 mg
Formula Weight 574.11
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Inosine 5′-Triphosphate Trisodium Salt Hydrate
TSCA No
Recommended Storage Refrigerator