missing translation for 'onlineSavingsMsg'
Learn More

Inosine 5'-Monophosphate Disodium Salt Hydrate 98.0+%, TCI America™

Catalog No. I003625G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I003625G 25 g
I00361G 1 g
I00365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I003625G Supplier TCI America Supplier No. I003625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4691-65-0
Molecular Formula C10H11N4Na2O8P
Molecular Weight (g/mol) 392.17
MDL Number MFCD00036201,MFCD00036201
InChI Key AANLCWYVVNBGEE-IDIVVRGQSA-L
Synonym disodium 5'-inosinate
PubChem CID 123134061
IUPAC Name disodium [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-6,9-dihydro-3H-purin-9-yl)oxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1NC=NC2=O

Specifications

Color White
Quantity 25 g
Formula Weight 392.17
Percent Purity ≥98.0% (HPLC,E)
Physical Form Crystalline Powder
Chemical Name or Material Inosine 5′-Monophosphate Disodium Salt Hydrate
EINECSNumber (8)-0276
RTECSNumber NM7519500
TSCA Yes