Learn More
Alkali Scientific Inoculating Needles, Rigid ABS (50 Sterile Peel Bags of 20 Needles), 1000/cs
Catalog No. 502335880
Encompass
These sterile, rigid ABS, disposable plastic needles are designed for use in quantitative procedures such as sampling, urine counts and serial dilutions, as well as for bacterial inoculation. Because they are pre-sterilized and designed for single use, there is no risk of cross contamination and no need for flaming. These inoculating needles are flexible enough for streaking, yet rigid enough for subsurface inoculation. The unique hexagonal shape of the shaft makes it easier to grip, orientate and guide through a smooth streaking process. The loop surfaces are extremely smooth with no rough edges and no flashing or burrs on the loop head. This results in the elimination of cutting or gouging of the agar surface. sterile, disposable and verifiably accurate, these inoculation needles provide an efficient, cost-effective and time-saving alternative to traditional metallic needles.
Catalog No. 502335880 Supplier Alkali Scientific Supplier No. M2030
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These sterile, rigid ABS, disposable plastic needles are designed for use in quantitative procedures such as sampling, urine counts and serial dilutions, as well as for bacterial inoculation. Because they are pre-sterilized and designed for single use, there is no risk of cross contamination and no need for flaming. These inoculating needles are flexible enough for streaking, yet rigid enough for subsurface inoculation. The unique hexagonal shape of the shaft makes it easier to grip, orientate and guide through a smooth streaking process. The loop surfaces are extremely smooth with no rough edges and no flashing or burrs on the loop head. This results in the elimination of cutting or gouging of the agar surface. sterile, disposable and verifiably accurate, these inoculation needles provide an efficient, cost-effective and time-saving alternative to traditional metallic needles.