missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Inoculating Nichrome Loops 8 in. Handle Braided/Twisted Wire Loop

Catalog No. 13820039
Change view
Click to view available options
Size:
2 mm
Includes:
Twisted Nichrome wire with aluminum handle
Twisted Nichrome wire with insulated handle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Includes
13820039 2 mm Twisted Nichrome wire with insulated handle
13820035 2 mm Twisted Nichrome wire with aluminum handle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13820039 Supplier Fisherbrand™ Supplier No. 10401140

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fisherbrand™ Innoculating loops are available in a variety of gauges, loop hole sizes, and wire types to accommodate an array of laboratory requirements.

Fisherbrand™ Nichrome/Aluminum Innoculating Loops consist of a nichrome wire loop mounted to an aluminum handle. The aluminum handle is available in either an insulated or non-insulated format.

  • Ideal for selecting, inoculating, or transferring microbiological samples
  • Cools rapidly after sterilization
  • Loops Slide along agar surfaces without cutting or tearing material
  • Nichrome wire available in single or twisted configuration
  • Passivated
  • 203 mm (8 in.) handles, available with insulation
  • Compatible with Gamma/ETO/Autoclavable sterilization methods

Certifications

ISO 9001-02, GMP, cGMP

Specifications

Material Nichrome
Length (Metric) 280 mm
Length (English) 11 in.
Size 2 mm
Color Silver
Product Type Inoculating Loop
For Use With (Equipment) Petri-Dish etc
Gauge 24
Includes Twisted Nichrome wire with insulated handle
Autoclavable Autoclavable
For Use With (Application) Ideal for selecting, inoculating, or transferring microbiological samples
Show More Show Less